Få mere at vide
Cyclohexanecarbohydrazide, TRC
Artikelnummer. 30354914
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354914 100 mg
30359916 500 mg
30363760 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30354914 Leverandør Toronto Research Chemicals Leverandørnr. TRCC987303100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38941-47-8
Molekylær formel C7H14N2O
Molekylvægt (g/mol) 142.19
IUPAC navn cyclohexanecarbohydrazide
SMIL NNC(=O)C1CCCCC1
Tekniske data
Kemisk navn eller materiale Cyclohexanecarbohydrazide
Mængde 100 mg
InChI formel InChI=1S/C7H14N2O/c8-9-7(10)6-4-2-1-3-5-6/h6H,1-5,8H2,(H,9,10)
Formel vægt 142.1106