missing translation for 'onlineSavingsMsg'
Få mere at vide

Cyclohexane-d11, CDN

Artikelnummer. 30312929
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312929 0.5 g 0.5g
30310160 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312929 Leverandør CDN Leverandørnr. CDND02950.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Cyclohexane-D11
CAS 1520-59-8
Molekylær formel C6 D11 H
Mængde 0.5 g
Synonym Cyclohexane-1,1,2,2,3,3,4,4,5,5,6-d11 (ACI), Cyclohexane-d11 (6 CI, 7 CI, 8 CI, 9 CI), Undecadeuteriocyclohexane, Undecadeuterocyclohexane, Cyclohexane-D11
InChI formel InChI=1 S/C6H12/c1-2-4-6-5-3-1/h1-6H2/i1D,2D2,3D2,4D2,5D2,6D2
SMIL [2 H]C1C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6-undecadeuteriocyclohexane
Molekylvægt (g/mol) 95.2273
Formel vægt 95.1629 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity
Vis mere Vis mindre
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.