Få mere at vide
Cyclohexanamine-D11, TRC
Artikelnummer. 30356139
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30356139 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30356139 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98455725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclohexanamine-D11
CAS 1219805-96-5
Molekylær formel C6 2H11 H2 N
Mængde 25 mg
Synonym Cyclohexan-1,2,2,3,3,4,4,5,5,6,6-d11-amine (ACI), Cyclohexanamine-D11, Cyclohexyl-d11-amine
InChI formel InChI=1S/C6H13N/c7-6-4-2-1-3-5-6/h6H,1-5,7H2/i1D2,2D2,3D2,4D2,5D2,6D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(N)C([2H])([2H])C1([2H])[2H]
IUPAC navn 1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexan-1-amine
Molekylvægt (g/mol) 110.24
Formel vægt 110.1738