Få mere at vide
Cycloguanil Embonate, Mikromol™
Beskrivelse
Cycloguanil Embonate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Cycloguanil Embonate |
| CAS | 609-78-9 |
| Analyt- eller komponentnavne | Cycloguanil Embonate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C23 H16 O6 . 2 C11 H14 Cl N5 |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 1-(4-chlorophenyl)-1,6-dihydro-6,6-dimethyl-1,3,5-triazine-2,4-diamine (1:2), 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 4,6-diamino-1-(p-chlorophenyl)-1,2-dihydro-2,2-dimethyl-s-triazine (1:2) (8CI), 1,3,5-Triazine-2,4-diamine, 1-(4-chlorophenyl)-1,6-dihydro-2,2-dimethyl-, 4,4'-methylenebis[3-methyl-2-naphthalenecarboxylate] (2:1) (9CI), s-Triazine, 4,6-diamino-1-(p-chlorophenyl)-1,2-dihydro-2,2-dimethyl-, 4,4'-methylenebis[3-hydroxy-2-naphthoate] (2:1) (8CI), 4,6-Diamino-1-(p-chlorophenyl)-1,2-dihydro-2,2-dimethyl-s-triazine compound (2:1) with 4,4'-methylenebis[3-hydroxy-2-naphthoic acid], 4,6-Diamino-1-(p-chlorophenyl)-1,2-dihydro-2,2-dimethyl-s-triazine pamoate, CI 501, Camolar, Cycloguanil embonate, Cycloguanil pamoate, Dihydrotriazine pamoate, NSC 77830, PAM-MR 807-23a |
| InChI formel | InChI=1S/C23H16O6.2C11H14ClN5/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;2*1-11(2)16-9(13)15-10(14)17(11)8-5-3-7(12)4-6-8/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2*3-6H,1-2H3,(H4,13,14,15,16) |
| SMIL | CC1(C)N=C(N)N=C(N)N1c2ccc(Cl)cc2.CC3(C)N=C(N)N=C(N)N3c4ccc(Cl)cc4.OC(=O)c5cc6ccccc6c(Cc7c(O)c(cc8ccccc78)C(=O)O)c5O |
| IUPAC navn | 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;1-(4-chlorophenyl)-6,6-dimethyl-1,3,5-triazine-2,4-diamine |
| Vis mere |