missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
MedChemExpress Cyclo(-RGDfK)
Specifications
Specifications
| Renhed | 98.0% |
| CAS | 161552-03-0 |
| Indhold og opbevaring | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| Til brug med (applikation) | Cancer-programmed cell death |
| Molekylvægt (g/mol) | 603.67 g/mol |
| Produkttype | Peptides |
| Mængde | 100 mg |
| Forsendelsestilstand | Room temperature |
| Opløselighed | DMSO : 125 mg/mL (207.07 mM∣Need ultrasonic) ∣H2O : 50 mg/mL (82.83 mM∣Need ultrasonic) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?