missing translation for 'onlineSavingsMsg'
Learn More

Cyclo(L-prolyl-L-valyl), TRC

Product Code. 30352684
Change view
Click to view available options
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30352684 5 mg
30359988 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30352684 Supplier Toronto Research Chemicals Supplier No. TRCC9898255MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2854-40-2
Molekylær formel C10H16N2O2
Molekylvægt (g/mol) 196.25
Synonym (3S-trans)-Hexahydro-3-(1-methylethyl)-pyrrolo[1,2-a]pyrazine-1,4-dione
IUPAC navn (3S,8aS)-3-isopropylhexahydropyrrolo[1,2-a]pyrazine-1,4-dione
SMIL O=C([C@@H](N1)C(C)C)N(CCC2)[C@]2([H])C1=O

Specifications

Kemisk navn eller materiale Cyclo(L-prolyl-L-valyl)
Mængde 5 mg
InChI formel InChI=1S/C10H16N2O2/c1-6(2)8-10(14)12-5-3-4-7(12)9(13)11-8/h6-8H,3-5H2,1-2H3,(H,11,13)/t7-,8-/m0/s1
Formel vægt 196.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.