Få mere at vide
Cyclo(L-Ala-L-Gly), TRC
Artikelnummer. 30360806
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30360806 1 g
1 options
Artikelnummer. 30360806 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9897701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclo(L-Ala-L-Gly)
CAS 4526-77-6
Molekylær formel C5H8N2O2
Mængde 1 g
Synonym (S)-3-Methyl-2,5-piperazinedione, L-3-Methyl-2,5-piperazinedione, Cyclo(L-alanylglycyl), Alanine, cyclic anhydride with glycine, Cyclo(Ala-Gly), Cyclo(Gly-L-Ala), Cyclo(alanylglycine), Cyclo(glycylalanine), N-Glycylalanine cyclic anhydride
InChI formel InChI=1S/C5H8N2O2/c1-3-5(9)6-2-4(8)7-3/h3H,2H2,1H3,(H,6,9)(H,7,8)/t3-/m0/s1
SMIL C[C@@H]1NC(=O)CNC1=O
IUPAC navn (3S)-3-methylpiperazine-2,5-dione
Molekylvægt (g/mol) 128.13
Formel vægt 128.0586