Få mere at vide
MedChemExpress Cyclo(his-pro)
Artikelnummer. 16414596
Encompass_Preferred
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
16414596 100 mg
1 options
Artikelnummer. 16414596 Leverandør MedChemExpress Leverandørnr. HY101402
MedChemExpress Cyclo(his-pro) (Cyclo(histidyl-proline)) is an orally active cyclic dipeptide structurally related to tyreotropin-releasing hormone. Cyclo(his-pro) could inhibit NF-κB nuclear accumulation. Cyclo(his-pro) can cross the brain-blood-barrier and affect diverse inflammatory and stress responses.
Tekniske data
CAS 53109-32-3
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Til brug med (applikation) Neuroscience-Neuromodulation
Molekylvægt (g/mol) 234.25 g/mol
Produkttype Natural Products
Mængde 100 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : ≥ 30 mg/mL (128.07 mM)
Synonym Cyclo(histidyl-proline)∣ Histidylproline diketopiperazine