Få mere at vide
Cyclizine-d4 Hydrochloride, TRC
Artikelnummer. 30368741
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30368741 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368741 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9878075MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclizine-d4 Hydrochloride
CAS 1246814-82-3
Molekylær formel C18H19D4ClN2
Mængde 5 mg
Synonym 1-(Diphenylmethyl)-4-methylpiperazine-d4 Hydrochloride, Marezine-d4 Hydrochloride, Marzine-d4, N-Benzhydryl-N'-methylpiperazine-d4 Monohydrochloride
InChI formel InChI=1S/C18H22N2.ClH/c1-19-12-14-20(15-13-19)18(16-8-4-2-5-9-16)17-10-6-3-7-11-17;/h2-11,18H,12-15H2,1H3;1H/i14D2,15D2;
SMIL [2H]C1(CN(CC(N1C(C2=CC=CC=C2)C3=CC=CC=C3)([2H])[2H])C)[2H].Cl
IUPAC navn 1-benzhydryl-2,2,6,6-tetradeuterio-4-methylpiperazine;hydrochloride
Molekylvægt (g/mol) 306.87