missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Cyclin B1 Monoclonal Antibody (GNS1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514319
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MA5-14319 targets Cyclin B1 in FACS, IF, IP, and WB applications and shows reactivity with Hamster, Human, and mouse samples. The MA5-14319 immunogen is human cyclin B1 protein.
Cyclins bind to and regulate the activity of the Cyclin dependent protein kinases (CDKs). Cyclin B1 or CCNB1 is a regulatory protein involved in mitosis. It is essential for the control of the cell cycle at the G2/M transition. Cyclin B1 complexes with p34 (cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. Cyclin B1 is not ubiquitinated during G2/M phase, resulting in its steady accumulation during G2 phase, followed by abrupt APC dependent destruction at the end of mitosis. Destruction of Cyclin B1 is required for cell cycle progression. Cyclin B1 is overexpressed in various cancers, including breast, prostate, and non-small cell lung cancer.
Tekniske data
| Cyclin B1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P14635, P24860, Q08301 | |
| Ccnb1 | |
| Human cyclin B1 protein | |
| 500 μL | |
| Primary | |
| Hamster, Human, Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| GNS1 | |
| Unconjugated | |
| Ccnb1 | |
| Ccn-2; CCNB; Ccnb1; Ccnb1-rs1; Ccnb1-rs13; Cycb; Cycb1; Cycb1-rs1; Cycb-4; Cycb-5; cyclin B; cyclin B1; cyclin B1, related sequence 1; cyclin B1, related sequence 13; G2/mitotic-specific cyclin B1; G2/mitotic-specific cyclin-B1; I79_013589; OTTHUMP00000221981; OTTHUMP00000223023 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100770842, 268697, 891 | |
| 4° C | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion