Få mere at vide
Cyclandelate, LoGiCal
Artikelnummer. 30459602
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459602 10 mg
1 options
Artikelnummer. 30459602 Leverandør LoGiCal Leverandørnr. LGCFOR0497.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclandelate
CAS 456-59-7
Analyt- eller komponentnavne Cyclandelate
Forsendelsestilstand Room Temperature
Molekylær formel C17 H24 O3
Certificeringer/compliance ISO 9001
Synonym Mandelic acid, 3,3,5-trimethylcyclohexyl ester (6CI,7CI,8CI), Cyclohexanol, 3,3,5-trimethyl-, mandelate (8CI), 3,3,5-Trimethylcyclohexyl mandelate, 3,5,5-Trimethylcyclohexanol, mandelic acid ester, 3,5,5-Trimethylcyclohexyl amygdalate, 3,5,5-Trimethylcyclohexyl mandelate, Arto-Espasmol, BS 572, Capilan, Ciclospasmol, Clandilon, Cyclandelate, Cyclergine, Cyclobral, Cyclolyt, Cyclomandol, Cyclospasmol, Dilatan, Natil, Novodil, Perebral, Saiclate, Sancyclan, Sepyron, Spasmione, Spasmocyclon, Benzeneacetic acid, alpha-hydroxy-, 3,3,5-trimethylcyclohexyl ester, 3,3,5-Trimethylcyclohexanol alpha-phenyl-alpha-hydroxyacetate
InChI formel InChI=1S/C17H24O3/c1-12-9-14(11-17(2,3)10-12)20-16(19)15(18)13-7-5-4-6-8-13/h4-8,12,14-15,18H,9-11H2,1-3H3
Løsningstype Neat
SMIL CC1CC(CC(C)(C)C1)OC(=O)C(O)c2ccccc2
IUPAC navn (3,3,5-trimethylcyclohexyl) 2-hydroxy-2-phenylacetate
Industry Type Clinical Testing
Molekylvægt (g/mol) 276.37
Mængde 10 mg
Formel vægt 276.1725 g/mol