Få mere at vide
Cyclamic Acid-d11, TRC
Artikelnummer. 30353114
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30353114 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353114 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9877221MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclamic Acid-d11
CAS 1215770-73-2
Molekylær formel C6 D11 H2 N O3 S
Mængde 1 mg
Synonym (1,2,2,3,3,4,4,5,5,6,6-Undecadeuteriocyclohexyl)sulfamic acid, Cyclamic acid D11 (cyclohexyl D11), Cyclamic acid-d11, N-(Cyclohexyl-d11)sulfamic acid, Cyclamate-d11, [2H11]Cyclohexylamidosulfuric acid, Cyclohexyl-d11-aminesulfonic acid, Cyclohexylsulfamic acid-d11
InChI formel InChI=1S/C6H13NO3S/c8-11(9,10)7-6-4-2-1-3-5-6/h6-7H,1-5H2,(H,8,9,10)/i1D2,2D2,3D2,4D2,5D2,6D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(NS(=O)(=O)O)C([2H])([2H])C1([2H])[2H]
IUPAC navn (1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)sulfamic acid
Molekylvægt (g/mol) 190.305
Formel vægt 190.131