Få mere at vide
Cyanomethyl 2,3,4,6-Tetra-O-acetyl-1-thio-Beta-D-galactopyranoside, TRC
Artikelnummer. 30365431
Skift visning
Mængde:
500 mg
1 g
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365431 2 g
30364171 1 g
30360515 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30365431 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9819502G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61145-33-3
Molekylær formel C16 H21 N O9 S
Molekylvægt (g/mol) 403.4
IUPAC navn [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(cyanomethylsulfanyl)oxan-2-yl]methyl acetate
SMIL CC(=O)OC[C@H]1O[C@@H](SCC#N)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C
Tekniske data
Kemisk navn eller materiale Cyanomethyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-galactopyranoside
Mængde 2 g
InChI formel InChI=1S/C16H21NO9S/c1-8(18)22-7-12-13(23-9(2)19)14(24-10(3)20)15(25-11(4)21)16(26-12)27-6-5-17/h12-16H,6-7H2,1-4H3/t12-,13+,14+,15-,16+/m1/s1
Formel vægt 403.0937