Få mere at vide
Cyanoguanidine-15N4, TRC
Artikelnummer. 30011250
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011250 25 mg
30010447 5 mg
30011243 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011250 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98135225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 63419-77-2
Molekylær formel C2 H4 15N4
Molekylvægt (g/mol) 88.0536
Synonym N-Cyanoguanidine-15N4, 1-Cyanoguanidine-15N4, 2-Cyanoguanidine-15N4, CGNA-15N4, Dicyandiamide-15N4, Dicyanodiamide-15N4, Didin-15N4, Epicure DICY-15N4
IUPAC navn 1-((^{15}N)azanylidynemethyl)guanidine
SMIL [15NH2]C(=[15NH])[15NH]C#[15N]
Tekniske data
Kemisk navn eller materiale Cyanoguanidine-15N4
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C2H4N4/c3-1-6-2(4)5/h(H4,4,5,6)/i3+1,4+1,5+1,6+1
Formel vægt 88.0317
Procent renhed >95%