Få mere at vide
Cyanocobalamin-d-carboxylic Acid, TRC
Artikelnummer. 30009192
Skift visning
Mængde:
1 mg
2.5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30009192 2.5 mg 2.5mg
30004368 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009192 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9536232.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38218-55-2
Molekylær formel C8 H8 Cl2 O2
Molekylvægt (g/mol) 207.054
Synonym 3'-Ester with 5,6-Dimethyl-1-alpha-D-ribofuranosylbenzimidazole Cobinic Acid-abceg-pentamide Cyanide Phosphate Inner Salt, 3'-Ester with 5,6-Dimethyl-1-alpha-D-ribofuranosyl-1H-benzimidazole Cobinic Acid-abceg-pentamide Cyanide Dihydrogen Phosphate (Ester) Inner Salt, 3'-Ester with (5,6-Dimethyl-1-alpha-D-ribofuranosyl-1H-benzimidazole-kN3) Cobinic Acid-abceg-pentamide Co-(cyano-kC)-Dihydrogen Phosphate (Ester) Inner Salt, CBC 226, Cobalamin-d-monoacid Cyanide
IUPAC navn 1-(2,4-dichlorophenyl)ethane-1,2-diol
SMIL OCC(O)c1ccc(Cl)cc1Cl
Tekniske data
Kemisk navn eller materiale Cyanocobalamin-d-carboxylic Acid
Mængde 2.5 mg
InChI formel InChI=1S/C8H8Cl2O2/c9-5-1-2-6(7(10)3-5)8(12)4-11/h1-3,8,11-12H,4H2
Formel vægt 205.99