Learn More
Macherey-Nagel™ Cyano Phases Polygosil™
Brand:  Macherey-Nagel™ 711390.100
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10669371
  • 10920.00 DKK / 100g
Estimated Shipment: 23-05-2024
to see stock.
Specifications
10 μm
Liquid Chromatography
Polygosil CN Phases
60 Å
100 g