missing translation for 'onlineSavingsMsg'
Learn More
Cyanide (CN-) @ 1000 mg/L in 0.1% NaOH, VHG™ Product Code.: 17989714

Cyanide (CN-) @ 1000 mg/L in 0.1% NaOH, VHG™

Product Code. p-200131996
500 mL, 500mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 17989714

Brand: VHG Labs VHGCN500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Cyanide (CN-) @ 1000 mg/L in 0.1% NaOH

Specifications

Quantity 500 mL
Solvent or Matrix Dilute NaOH
CAS 57-12-5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyanide (CN-) @ 1000 mg/L in 0.1% NaOH, VHG™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.