Få mere at vide
Cyamemazine-d6, TRC
Artikelnummer. 30364399
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30364399 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30364399 Leverandør Toronto Research Chemicals Leverandørnr. TRCC95150210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyamemazine-d6
CAS 1216608-24-0
Renhedsgrad noter HPLC
Molekylær formel C19H15D6N3S
Mængde 10 mg
Synonym Cyamemazine D6
InChI formel InChI=1S/C19H21N3S/c1-14(12-21(2)3)13-22-16-6-4-5-7-18(16)23-19-9-8-15(11-20)10-17(19)22/h4-10,14H,12-13H2,1-3H3/i2D3,3D3
SMIL [2H]C([2H])([2H])N(CC(C)CN1c2ccccc2Sc3ccc(cc13)C#N)C([2H])([2H])[2H]
IUPAC navn 10-[3-[bis(trideuteriomethyl)amino]-2-methylpropyl]phenothiazine-2-carbonitrile
Molekylvægt (g/mol) 329.49
Procent renhed >95