missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL8/IL-8 Mouse anti-Bovine, Canine, Clone: MT8H6, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18279-1000UG
Additional Details : Weight : 0.00970kg
Description
CXCL8/IL-8 Monoclonal antibody specifically detects CXCL8/IL-8 in Bovine, Canine samples. It is validated for ELISASpecifications
CXCL8/IL-8 | |
Monoclonal | |
0.5 mg/ml | |
0.2 um filtered PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Bovine, Canine | |
Purified |
ELISA | |
MT8H6 | |
Unconjugated | |
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor | |
Recombinant human CXCL8/ IL-8 | |
1000 μg | |
Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
3576 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |