missing translation for 'onlineSavingsMsg'
Learn More
CX-6258, MedChemExpress Product Code.: 16487458

CX-6258, MedChemExpress

Product Code. 16487458
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487458

Brand: Medchem Express HY18095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CX-6258 is a potent and kinase selective pan-Pim kinases inhibitor, with IC50s of 5 nM, 25 nM and 16 nM for Pim-1, Pim-2 and Pim-3, respectively.

CX-6258 is a potent and kinase selective pan-Pim kinases inhibitor, with IC50s of 5 nM, 25 nM and 16 nM for Pim-1, Pim-2 and Pim-3, respectively.

Specifications

Chemical Name or Material CX-6258
CAS 1202916-90-2
Color Orange
Purity Grade Notes Research
Molecular Formula C26H24ClN3O3
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (108.24 mM)
SMILES O=C1NC2=CC=C(Cl)C=C2/C1=C\C3=CC=C(C4=CC=CC(C(N5CCCN(C)CC5)=O)=C4)O3
Molecular Weight (g/mol) 461.94
Formula Weight 461.94
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
CX-6258, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.