Få mere at vide
R&D Systems™ CX-4945
Beskrivelse
CX-4945 can be used, in combination with other small molecules, in fast chemical reprogramming protocols for the induction of human somatic cells and highly efficient generation of pluripotent stem cells (hCiPS) in a minimum of 16 days with robust repeatability and reliability.
CX-4945 has antiproliferative effects in vitro as it attenuates PI3K/Akt signalling, causes cell-cycle arrest and selectively induces apoptosis in cancer cells. In models of angiogenesis, CX-4945 inhibits human umbilical vein endothelial cell migration, tube formation, and blocks CK2-dependent hypoxia-induced factor 1 alpha (HIF-1α) transcription in cancer cells.
In vivo, CX-4945 displays robust antitumor activity with concomitant reduction of the mechanism-based biomarker phospho-p21 (T145) (murine xenograft model). CX-4945 is orally bioavailable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Sodium 5-[(3-Chlorophenyl)amino]benzo[c]-2,6-naphthyridine-8-carboxylate |
| CAS | 1309357-15-0 |
| Mængde | 10 mg |
| Produktlinje | Tocris |
| Mål | Casein Kinase 2 Inhibitors |
| Molekylær formel | C19H11ClN3O2Na |
| Renhed | 98% |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.