Learn More
Macherey-Nagel™ CW Deactivated Capillary Column
Brand: Macherey-Nagel™ 723105.25
Additional Details : Weight : 0.01000kg
Specifications
0.25 mm | |
0.4 mm | |
250°C | |
1 Each |
Deactivated Capillary Column | |
Preparation of Capillary Columns Analysis | |
25 m |