Learn More
CVT-12012, MedChemExpress
Product Code. 16475276
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475276 100 mg 100mg
16467097 5 mg 5mg
16453056 25 mg 25mg
16450697 50 mg 50mg
16414378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16475276 Supplier Medchem Express Supplier No. HY11034
 Restricted Item
Forventet forsendelse: 30-11-2026
MedChemExpress CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively.
CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively.
Specifications
Chemical Name or Material CVT-12012
Physical Form Powder
CAS 1018675-35-8
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H21F3N4O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 125 mg/mL (287.75 mM; Need ultrasonic)
SMILES O=C(NCCN1C(C(C)=NC2=C1C=C(NCC3=CC=CC(C(F)(F)F)=C3)C=C2)=O)CO
Molecular Weight (g/mol) 434.41
Formula Weight 434.41
Percent Purity 99.16%
Grade Research