Learn More
Cutaneous Lymphocyte Antigen (CLA) Monoclonal Antibody (HECA-452), eFluor™ 660, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 50-9857-82
Additional Details : Weight : 0.09500kg
Description
This cell surface glycoprotein is most highly expressed on T cells in the skin. Nevertheless, CLA can also be detected on some peripheral T and NK cells, as well as monocytes, neutrophils and CD1a+ dendritic cells. Studies have determined that this antibody recognizes sialofucosylated glycans and sialyl Lewis x (sLex). CLA, which binds E-selectin and endothelial cell-leukocyte adhesion molecule 1 (ELAM-1), is involved in T lymphocyte infiltration and adhesion at cutaneous sites of inflammation.
Cutaneous Lymphocyte Antigen (CLA) Monoclonal antibody specifically detects Cutaneous Lymphocyte Antigen (CLA) in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Cutaneous Lymphocyte Antigen (CLA) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
100 μg | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
HECA-452 | |
eFluor 660 | |
CLA | |
Affinity chromatography | |
RUO | |
Human | |
Antibody | |
IgM κ |
For Research Use Only.