Få mere at vide
Custom Primer
Artikelnummer. 10657173
Artikelnummer. 10657173 Leverandør Invitrogen Leverandørnr. 56830
 Begrænset vare
Forventet forsendelse: 13-11-2026
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).