Learn More
Custom Primer
Product Code. 10347833
Product Code. 10347833 Supplier Invitrogen Supplier No. 11751021
 Restricted Item
Forventet forsendelse: 11-12-2026
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).