Få mere at vide
Cuscohygrine (Mixture of Diastereomers), TRC
Artikelnummer. 30011612
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011612 100 mg
30011512 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011612 Leverandør Toronto Research Chemicals Leverandørnr. TRCC845000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 454-14-8
Molekylær formel C13 H24 N2 O
Molekylvægt (g/mol) 224.34
Synonym 2-Propanone, 1,3-bis(1-methyl-2-pyrrolidinyl)-, (R*,S*)-, Cuscohygrine (6CI,7CI,8CI), rel-1-[(2R)-1-Methyl-2-pyrrolidinyl]-3-[(2S)-1-methyl-2-pyrrolidinyl]-2-propanone, Bellaradin, Bellaradine, Cuscohygrine, meso-, Cuskhygrin, Cuskhygrine, meso-Cuscohygrine, 2-Propanone, 1-[(2R)-1-methyl-2-pyrrolidinyl]-3-[(2S)-1-methyl-2-pyrrolidinyl]-, rel-
IUPAC navn 1-[(2S)-1-methylpyrrolidin-2-yl]-3-[(2R)-1-methylpyrrolidin-2-yl]propan-2-one
SMIL CN1CCC[C@H]1CC(=O)C[C@H]2CCCN2C
Tekniske data
Kemisk navn eller materiale Cuscohygrine (Mixture of Diastereomers)
Mængde 100 mg
InChI formel InChI=1S/C13H24N2O/c1-14-7-3-5-11(14)9-13(16)10-12-6-4-8-15(12)2/h11-12H,3-10H2,1-2H3/t11-,12+
Formel vægt 224.1889