Få mere at vide
Cuscohygrine-d6 (Mixture of Diastereomers), TRC
Artikelnummer. 30354700
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30354700 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354700 Leverandør Toronto Research Chemicals Leverandørnr. TRCC8450021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cuscohygrine N,N-dimethyl-d6 (mixture of diastereomers)
CAS 1292907-65-3
Molekylær formel C13 D6 H18 N2 O
Mængde 1 mg
Synonym rel-1-[(2R)-1-(Methyl-d3)-2-pyrrolidinyl]-3-[(2S)-1-(methyl-d3)-2-pyrrolidinyl]-2-propanone, (R*,S*)-1,3-Bis[1-(methyl-d3)-2-pyrrolidinyl]-2-propanone, Bellaradin-d6, Bellaradine-d6, Cuskhygrin-d6, Cuskhygrine-d6, meso-Cuscohygrine-d6
InChI formel InChI=1S/C13H24N2O/c1-14-7-3-5-11(14)9-13(16)10-12-6-4-8-15(12)2/h11-12H,3-10H2,1-2H3/t11-,12+/i1D3,2D3
SMIL [2H]C([2H])([2H])N1CCC[C@@H]1CC(=O)C[C@@H]2CCCN2C([2H])([2H])[2H]
IUPAC navn 1-[(2S)-1-(trideuteriomethyl)pyrrolidin-2-yl]-3-[(2R)-1-(trideuteriomethyl)pyrrolidin-2-yl]propan-2-one
Molekylvægt (g/mol) 230.38
Formel vægt 230.2265