missing translation for 'onlineSavingsMsg'
Få mere at vide

Curcumin-d6 (dimethoxy-d6), CDN

Artikelnummer. 30321990
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.005 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321990 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321990 Leverandør CDN Leverandørnr. CDND79160.005G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Curcumin-d6
CAS 1246833-26-0
Molekylær formel C21 2H6 H14 O6
Mængde 0.005 g
InChI formel InChI=1 S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-12,24-25 H,13H2,1-2H3/b7-3+,8-4+/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])Oc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)c(OC([2 H])([2 H])[2 H])c2)ccc1O
IUPAC navn (1 E,6 E)-1,7-bis[4-hydroxy-3-(trideuteriomethoxy)phenyl]hepta-1,6-diene-3,5-dione
Molekylvægt (g/mol) 374.42
Formel vægt 374.1636 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.