Få mere at vide
Cumyl Bromoacetate, TRC
Artikelnummer. 30356287
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356287 5 g
30352820 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30356287 Leverandør Toronto Research Chemicals Leverandørnr. TRCC8347055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 153698-47-6
Molekylær formel C11H13BrO2
Molekylvægt (g/mol) 257.12
Synonym 2-Bromoacetic Acid 1-Methyl-1-phenylethyl Ester, alpha-Cumyl Bromoacetate
IUPAC navn 2-phenylpropan-2-yl 2-bromoacetate
SMIL CC(C)(OC(=O)CBr)c1ccccc1
Tekniske data
Kemisk navn eller materiale Cumyl Bromoacetate
Mængde 5 g
InChI formel InChI=1S/C11H13BrO2/c1-11(2,14-10(13)8-12)9-6-4-3-5-7-9/h3-7H,8H2,1-2H3
Formel vægt 256.0099