missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Cullin 5 Recombinant Rabbit Monoclonal Antibody (K01_1P96)
GREENER_CHOICE

Artikelnummer. p-200114762
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μL
100 μL
Pakningsstørrelse:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30826972 100 μL 100µL
30825118 50 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30826972 Leverandør Invitrogen™ Leverandørnr. MA563553

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

Cullin 5 Recombinant Monoclonal Antibody for Western Blot

CUL5 is a core component of multiple SCF-like ECS (Elongin-Cullin 2/5-SOCS-box protein) E3 ubiquitin-protein ligase complexes, which mediate the ubiquitination and subsequent proteasomal degradation of target proteins. As a scaffold protein may contribute to catalysis through positioning of the substrate and the ubiquitin-conjugating enzyme. The functional specificity of the E3 ubiquitin-protein ligase complex depends on the variable substrate recognition component. ECS(SOCS1) seems to direct ubiquitination of JAk2. It seems to be involved poteosomal degradation of p53/TP53 stimulated by adenovirus E1B-55 kDa protein and may form a cell surface vasopressin receptor.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Cullin 5
Applikationer Western Blot
Klassifikation Recombinant Monoclonal
Klon K01_1P96
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene CUL5
Genadgangsnr. Q93034, Q9D5V5, Q9JJ31
Gene Alias 4921514I20Rik; 8430423K24Rik; AI852817; C030032G03Rik; C330021I08Rik; CUL5; CUL-5; cullin 5; cullin-5; Cullin-5 (vasopressin-activated calcium-mobilizing receptor-1); Vacm1; VACM-1; vasopressin-activated calcium-mobilizing receptor 1; vasopressin-activated calcium-mobilizing receptor protein; Vasopressin-activated calcium-mobilizing receptor-1
Gensymboler CUL5
Værtsarter Rabbit
Immunogen Recombinant protein of human Cullin 5.
Oprensningsmetode Protein A/G
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 64624, 75717, 8065
Målarter Human, Mouse, Rat
Indhold og opbevaring Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.