missing translation for 'onlineSavingsMsg'
Learn More

Cucumopine, TRC

Product Code. 30357345
Change view
Click to view available options
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30357345 500 mg
30349201 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357345 Supplier Toronto Research Chemicals Supplier No. TRCC833500500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Cucumopine
CAS 110342-24-0
Molekylær formel C11 H13 N3 O6
Mængde 500 mg
Synonym (4S,6S)-4-(2-carboxyethyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine-4,6-dicarboxylic Acid, 4-Carboxy-4-(2-carboxyethyl)spinacine, (4S-trans)-4-(2-carboxyethyl)-4,5,6,7-tetrahydro-1H-Imidazo[4,5-c]pyridine-4,6-dicarboxylic Acid
InChI formel InChI=1S/C11H13N3O6/c15-7(16)1-2-11(10(19)20)8-5(12-4-13-8)3-6(14-11)9(17)18/h4,6,14H,1-3H2,(H,12,13)(H,15,16)(H,17,18)(H,19,20)/t6-,11-/m0/s1
SMIL OC(=O)CC[C@@]1(N[C@@H](Cc2nc[nH]c12)C(=O)O)C(=O)O
IUPAC navn (4S,6S)-4-(2-carboxyethyl)-3,5,6,7-tetrahydroimidazo[4,5-c]pyridine-4,6-dicarboxylic acid
Molekylvægt (g/mol) 283.24
Formel vægt 283.0804
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.