Learn More
Sartorius Cubis™ II Precision (3-place) Balance, MCE User Interface
Cubis™ II Precision (3-Place) Balance features fully customizable hardware, software, and connectivity
Brand: Sartorius MCE1203S-2S00-U
59299.76 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Monolithical weighing system
- Integrated climate sensors
- Individual sample holders
- Error-free operation
- Individual QApp workflows
- Motorized auto-leveling
- Full end-to-end data integrity
- Integrated audit-trail
- State-of-the-art user management
- Excellent service support
- Integrated status center
- Integrated service functions
- Preventive maintenance on accredited standards
- TFT touch screen operation with readable display and easy to operate user interface
- Software Factory installed basic set of essential weighing applications
- Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Services
10-year warranty
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port | |
TFT Touchscreen | |
Balance, Accessories, Power supply, Manual, Factory test report | |
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim | |
±0.5 mg | |
Precision Balance | |
425 mm | |
122/284/373 mm | |
120/240 V | |
50/60 Hz (± 5%) |
1.2 kg | |
1 mg | |
±1 mg | |
Draft shield U: Manual glass | |
1 sec | |
10 years | |
240 mm | |
5.9/7.5/9.4/10.2 kg | |
100/240 V | |
Class I |
Product Content Correction
Product Title
Sartorius Cubis™ II Precision (3-place) Balance, MCE User Interface > MCE user interface; Capacity: 1,200 g ; Draft shield U: Manual glass
Spot an opportunity for improvement?Share a Content Correction