Learn More
Sartorius Cubis™ II Precision (3-place) Balance, MCE User Interface
BIOPHARMAGREENER_CHOICE
Cubis™ II Precision (3-Place) Balance features fully customizable hardware, software, and connectivity
Brand:  Sartorius MCE2203SR-S00
57571.89 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16223262
  • 76725.00 DKK / 1 unit
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Services
10-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
±0.5 mg
Precision Balance
425 mm
122/284/373 mm
120/240 V
50/60 Hz (± 5%)
2.2 kg
1 mg
±2 mg
Draft shield R: Stainless steel frame
1 sec
10 years
122 mm
5.9/7.5/9.4/10.2 kg
100/240 V
Class I
Videos
  • Sartorius - Cubis analytical balances
    Offers the optimal basis for users who need to weigh with the highest accuracy, but have lower needs in terms of complexity of a weighing process and in data management.
    Video Player is loading.
    Current Time 0:00
    Duration 1:26
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:26
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Sartorius Cubis analytical balances
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius Cubis™ II Precision (3-place) Balance, MCE User Interface > MCE user interface; Capacity: 2,200 g ; Draft shield R: Stainless steel frame; QP99 software