Få mere at vide
Tocris Bioscience™ CU-T12-9
Potent TLR1/2 agonist
Brand: Tocris Bioscience™ 5414/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CU-T12-9 is a potent TLR1/2 agonist (EC50 = 52.9 nM). Activates the NF-κB pathway and upregulates proinflammatory cytokines in vitro. Also enhances TLR1 and TLR2 dimerization.Tekniske data
| CU-T12-9 | |
| N-Methyl-4-nitro-2-(4-(4-trifluoromethyl)phenyl)-1H-imidazol-1-yl)aniline | |
| Soluble to 100mM in DMSO and to 20mM in ethanol | |
| Bioactive Small Molecule |
| 1821387-73-8 | |
| 50 mg | |
| Solid |
Produkttitel
Ser du en mulighed for forbedring?
Research Use Only. Not For Human or Veterinary Use