Learn More
CU-CPT22, MedChemExpress
MedChemExpress CU-CPT22 is a potent protein complex of toll-like receptor 1 and 2 (TLR1/2) inhibitor, and competes with the synthetic triacylated lipoprotein (Pam3CSK4) binding to TLR1/2 with a Ki of 0.41 μM. CU-CPT22 blocks Pam3CSK4-induced TLR1/2 activation with an IC50 of 0.58 μM.

1349.54 DKK - 4370.91 DKK
Specifications
Chemical Name or Material | CU-CPT22 |
---|---|
CAS | 1416324-85-0 |
Color | Orange |
Purity Grade Notes | Research |
Molecular Formula | C19H22O7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16466917
![]() |
Medchem Express
HY-108471 |
5 mg |
1349.54 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16404208
![]() |
Medchem Express
HY-108471 |
10 mg |
2286.17 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16402996
![]() |
Medchem Express
HY-108471 |
25 mg |
4370.91 DKK
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CU-CPT22 is a potent protein complex of toll-like receptor 1 and 2 (TLR1/2) inhibitor, and competes with the synthetic triacylated lipoprotein (Pam3CSK4) binding to TLR1/2 with a Ki of 0.41 μM. CU-CPT22 blocks Pam3CSK4-induced TLR1/2 activation with an IC50 of 0.58 μM.Specifications
CU-CPT22 | |
Orange | |
C19H22O7 | |
O=C(C(C=C1O)=CC2=CC(OC)=C(O)C(O)=C2C1=O)OCCCCCC | |
362.37 | |
Research | |
Cancer-programmed cell death |
1416324-85-0 | |
Research | |
DMSO : ≥ 125 mg/mL (344.95 mM) | |
362.37 | |
98.61% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.