missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ CU CPT 9a
GREENER_CHOICE

Artikelnummer. 18737766
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18737766 25 mg
18729006 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18737766 Leverandør Tocris Bioscience™ Leverandørnr. 6479/25

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Potent TLR8 inhibitor

CU CPT 9a is a potent TLR8 inhibitor (IC50 = 0.5 nM and Kd = 21 nM). Stabilizes the TLR8 dimer in its resting state, preventing activation. Suppresses TLR8-mediated proinflammatory signaling in various cell lines and human primary cells.
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Methyl-4-(7-methoxy-4-quinolinyl)phenol
CAS 2165340-32-7
Mængde 25 mg
Molekylær formel C17H15NO2
Renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.