Få mere at vide
Tocris Bioscience™ CU-76
Beskrivelse
CU-76 is a selective cyclic GMP-AMP synthase (cGAS) inhibitor (IC50 = 0.24 μM). It specifically inhibits cGAS-STING pathway and has no effect on other nucleic acid sensing pathways such as the RIG-I-MAVS or Toll-like receptor pathways. CU-76 reduces DNA-induced IFN regulatory factor 3 (IRF3) dimerization in a dose-dependent manner, and it decreases production of IFN-β in THP-1 cells stimulated with interferon-stimulatory DNA.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl 4-amino-6-[(3,5-difluoro-4-iodophenyl)amino]-1,3,5-triazine-2-carboxylate |
| CAS | 2400954-58-5 |
| Mængde | 2 mg |
| Mål | STING/TMEM173 |
| Molekylær formel | C11H8F2IN5O2 |
| Renhed | 0.98 |