Få mere at vide
CTX1, MedChemExpress
Artikelnummer. 16484106
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
Pakningsstørrelse:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16484106 25 mg 25mg
16472648 5 mg -
16460449 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16484106 Leverandør Medchem Express Leverandørnr. HYU00442
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
MedChemExpress CTX1 is a p53 activator that overcomes HdmX-mediated p53 repression. CTX1 exhibits potent anti-cancer activity in a mouse acute myeloid leukemia (AML) model system.
CTX1 is a p53 activator that overcomes HdmX-mediated p53 repression. CTX1 exhibits potent anti-cancer activity in a mouse acute myeloid leukemia (AML) model system.
Tekniske data
Kemisk navn eller materiale CTX1
CAS 501935-96-2
Farve Reddish Brown
Renhedsgrad noter Research
Molekylær formel C14H10N4
Mængde 25 mg
Opløselighedsinformation DMSO : 12.5 mg/mL (53.36 mM; ultrasonic and warming and heat to 60°C)
SMIL N#CC1=C(C=CC(N)=C2)C2=NC3=CC(N)=CC=C31
Molekylvægt (g/mol) 234.26
Formel vægt 234.26
Procent renhed 96.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death