Learn More
CTU2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA588908
Description
Immunogen sequence: HVLGAGGGPG PTQGEEQPPQ PPLDPQNLAR PPAPAQTEAL SQLFCSVRTL TAKEELLQTL RTHLILHMAR AHGYSKVMTG DSCTRLAIKL MTNLALGRGA FLAWDTGFSD ERHGDVVVVR PMRDHTLKEV AFYNRLFSVP SVFTPAVDTK APEKASIHRL MEAFILRLQT QFPSTVSTVY RTSEKLVKGP RDGPAAGDSG PRCLLCMCAL DVDAADSATA FGAQTSSRLS QMQSPIPLTE TRTPPGPCCS PGVGWAQRCG QGACRREDPQ ACIEEQLCYS CRVNMKDLPS LDPLPPYILA EAQLRTQRAW GLQEIRDCLI EDSDDEAGQS; Positive Samples: A-431, Jurkat; Cellular Location: Cytoplasm
Cytoplasmic tRNA 2-thiolation protein 2 (CUT2), also named as C16orf84 or NCS2, is a 515 amino acid protein, which belongs to the CUT2 family. C16orf84 localizes in the cytoplasm and Plays a central role in 2-thiolation of mcm5S2U at tRNA wobble positions of tRNA(Lys), tRNA(Glu) and tRNA(Gln). C16orf84 may act by forming a heterodimer with CTU1/ATPBD3 that ligates sulfur from thio-carboxylated URM1 onto the uridine of tRNAs at wobble position.
Specifications
CTU2 | |
Polyclonal | |
Unconjugated | |
CTU2 | |
2310061F22Rik, C16orf84, Ctu2, Cytoplasmic tRNA 2-thiolation protein 2, cytoplasmic tRNA 2-thiolation protein 2 {ECO:0000255, cytosolic thiouridylase subunit 2, cytosolic thiouridylase subunit 2 homolog, cytosolic thiouridylase subunit 2 homolog (S. pombe), HAMAP-Rule:MF_03054}, Ncs2, UPF0432 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
348180 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.72 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
Q2VPK5 | |
CTU2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 186-515 of human CTU2 (NP_0010127771) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.