missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CTSS (Human) Recombinant Protein
Human CTSS full-length ORF ( AAH02642, 1 a.a. - 331 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001520-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKRLVCVLLVCSSAVAQLHKDPTLDHHWHLWKKTYGKQYKEKNEEAVRRLIWEKNLKFVMLHNLEHSMGMHSYDLGMNHLGDMTSEEVMSLMSSLRVPSQWQRNITYKSNPNWILPDSVDWREKGCVTEVKYQGSCGACWAFSAVGALEAQLKLKTGKLVSLSAQNLVDCSTEKYGNKGCNGGFMTTAFQYIIDNKGIDSDASYPYKAMDQKCQYDSKYRAATCSKYTELPYGREDVLKEAVANKGPVSVGVDARHPSFFLYRSGVYYEPSCTQNVNHGVLVVGYGDLNGKEYWLVKNSWGHNFGEEGYIRMARNKGNHCGIASFPSYPEI
Specifications
AAH02642 | |
cathepsin S | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTSS | |
GST |
1520 | |
Wheat germ expression system | |
25 μg | |
MGC3886 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |