missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ CSTF3 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15574791
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μg
Pakningsstørrelse:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15574791 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15574791 Leverandør Invitrogen™ Leverandørnr. PA537856

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Goat Polyclonal Antibody

This antibody is predicted to react with mouse, rat and dog based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

The protein encoded by this gene is one of three (including CSTF1 and CSTF2) cleavage stimulation factors that combine to form the cleavage stimulation factor complex (CSTF). This complex is involved in the polyadenylation and 3' end cleavage of pre-mRNAs. The encoded protein functions as a homodimer and interacts directly with both CSTF1 and CSTF2 in the CSTF complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen CSTF3
Applikationer Western Blot
Klassifikation Polyclonal
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CSTF3
Genadgangsnr. Q12996
Gene Alias 4732468G05Rik; C79532; CF-1 77 kDa subunit; cleavage stimulation factor 77 kDa subunit; Cleavage stimulation factor subunit 3; cleavage stimulation factor subunit 3, isoform 1; cleavage stimulation factor, 3' pre-RNA, subunit 3; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kD; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa; CSTF 77 kDa subunit; CSTF3; CSTF-77; MGC117398; MGC43001; MGC75122
Gensymboler CSTF3
Værtsarter Goat
Immunogen Peptide with sequence C-RETKGKLPSYKE, from the internal region of CSTF3 (aa 109-121).
Oprensningsmetode Ammonium sulfate precipitation
Mængde 100 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 1479
Målarter Human
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.