missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ CSTF2T Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15585082
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15585082 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15585082 Leverandør Invitrogen™ Leverandørnr. PA541856

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Peptide sequence: MRGPVPSSRG PMTGGIQGPG PINIGAGGPP QGPRQVPGIS GVGNPGAGMQ Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 85%; Rabbit: 91%; Rat: 85%.

CSTF2T may play a significant role in AAUAAA-independent mRNA polyadenylation in germ cells. It is directly involved in the binding to pre-mRNAs.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen CSTF2T
Applikationer Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering PBS with 2% sucrose and 0.09% sodium azide
Gene CSTF2T
Genadgangsnr. Q9H0L4
Gene Alias 64kDa; C77975; CF-1 64 kDa subunit tau; CF-1 64 kDa subunit tau variant; CF-1 64 kDa subunit, tau; cleavage stimulation factor 64 kDa subunit tau; Cleavage stimulation factor 64 kDa subunit tau variant; cleavage stimulation factor 64 kDa subunit, tau; cleavage stimulation factor subunit 2 tau; cleavage stimulation factor subunit 2 tau variant; cleavage stimulation factor subunit 2, tau variant; cleavage stimulation factor, 3' pre-RNA subunit 2, 64 kDa, tau; cleavage stimulation factor, 3' pre-RNA subunit 2, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau variant; cleavage stimulation factor, 3' pre-RNA, subunit 2, tau; CSTF 64 kDa subunit tau; CSTF 64 kDa subunit tau variant; CSTF 64 kDa subunit, tau; CSTF2T; CstF-64T; Kiaa0689; TauCstF-64; tCstF-64; Unknown (protein for MGC:129118)
Gensymboler CSTF2T
Værtsarter Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CSTF2T (aa 460-509).
Oprensningsmetode Affinity Chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 23283
Målarter Human
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.