missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ CSTF2T Polyclonal Antibody

Beskrivelse
Antibody detects endogenous levels of total CSTF2T.
CSTF2T may play a significant role in AAUAAA-independent mRNA polyadenylation in germ cells. It is directly involved in the binding to pre-mRNAs.
Tekniske data
Tekniske data
| Antigen | CSTF2T |
| Applikationer | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CSTF2T |
| Genadgangsnr. | Q8C7E9, Q9H0L4 |
| Gene Alias | 64kDa; C77975; CF-1 64 kDa subunit tau; CF-1 64 kDa subunit tau variant; CF-1 64 kDa subunit, tau; cleavage stimulation factor 64 kDa subunit tau; Cleavage stimulation factor 64 kDa subunit tau variant; cleavage stimulation factor 64 kDa subunit, tau; cleavage stimulation factor subunit 2 tau; cleavage stimulation factor subunit 2 tau variant; cleavage stimulation factor subunit 2, tau variant; cleavage stimulation factor, 3' pre-RNA subunit 2, 64 kDa, tau; cleavage stimulation factor, 3' pre-RNA subunit 2, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau variant; cleavage stimulation factor, 3' pre-RNA, subunit 2, tau; CSTF 64 kDa subunit tau; CSTF 64 kDa subunit tau variant; CSTF 64 kDa subunit, tau; CSTF2T; CstF-64T; Kiaa0689; TauCstF-64; tCstF-64; Unknown (protein for MGC:129118) |
| Gensymboler | CSTF2T |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?