missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
Antibody detects endogenous levels of total CSTF2.
CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.
Tekniske data
Tekniske data
| Antigen | CSTF2 |
| Applikationer | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CSTF2 |
| Genadgangsnr. | P33240, Q8BIQ5 |
| Gene Alias | alphaCstF-64 variant 4; betaCstF-64 variant 1; betaCstF-64 variant 2; betaCstF-64 variant 3; C630034J23Rik; CF-1 64 kDa subunit; cleavage stimulation factor 64 kDa subunit; Cleavage stimulation factor subunit 2; cleavage stimulation factor, 3' pre-RNA subunit 2; cleavage stimulation factor, 3' pre-RNA subunit 2, 64 kDa; cleavage stimulation factor, 3' pre-RNA, subunit 2; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kD; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa; CstF 64; CSTF 64 kDa subunit; Cstf2; Cstf64; CstF-64; mir-3112 |
| Gensymboler | CSTF2 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?
