missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CSTF1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17237823 Shop All Thermo Scientific Products
Change view
Click to view available options
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17237823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17237823 Supplier Invitrogen™ Supplier No. PA5115058

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CSTF1.

This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSTF1
Applikationer Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide
Gene CSTF1
Genadgangsnr. Q05048, Q5BJQ6, Q99LC2
Gene Alias 1700057K18Rik; AI788832; CF-1 50 kDa subunit; cleavage stimulation factor 50 kDa subunit; Cleavage stimulation factor subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa; CSTF 50 kDa subunit; Cstf1; CstF50; CstF-50; CstFp50
Gensymboler CSTF1
Værtsarter Rabbit
Immunogen A synthesized peptide derived from human CSTF1(Accession Q05048), corresponding to amino acid residues M1-G33.
Oprensningsmetode Affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 1477, 311670, 67337
Målarter Human, Mouse, Rat
Indhold og opbevaring -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.