missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSRP1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99738AF594
Additional Details : Weight : 0.00970kg
Description
CSRP1 Polyclonal specifically detects CSRP1 in Human samples. It is validated for Western Blot, ELISA.Specifications
CSRP1 | |
Polyclonal | |
Western Blot, ELISA | |
CRP, CRP1, CSRPCYRP, cysteine and glycine-rich protein 1, Cysteine-rich protein 1, D1S181E, DKFZp686M148, LIM-domain protein | |
Produced in rabbits immunized with purified, recombinant CSRP1 (NP_004069.1; Met 1-Glu 193). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
1465 | |
Store at 4°C in the dark. | |
IgG |