missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CSNK1E (CK1 epsilon) [R178C], GST Tag Recombinant Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
Description
CSNK1E serine/threonine kinase is a member of the CK1 family which plays an important role in diverse cell processes, including DNA replication and repair. CSNK1E is a regulator of Yes-associated protein (YAP) transcription coactivator, which is a key regulator of organ size and a candidate human oncogene. CSNK1E is activated by CCK2R and then phosphorylates PKD2 at Ser244. Phosphorylation of PKD2 leads to its nuclear accumulation and efficient phosphorylation of nuclear PKD2 substrates in human gastric cancer cells. CSNK1E can phosphorylate topoisomerase (topo) II alpha at serine-1106 and this regulates the enzyme activity and sensitivity to topo II-targeted drugs.

Specifications
Specifications
Accession Number | P49674 |
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
Gene ID (Entrez) | 1454 |
Molecular Weight (g/mol) | 73.6 kDa |
Name | Human CSNK1E (CK1 epsilon) [R178C], GST Tag |
Quantity | 100 μg |
Gene Alias | CK1 |
Common Name | CK1 epsilon (R178C Mutant) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human CSNK1E (CK1 epsilon) [R178C], GST Tag Recombinant Protein >
Spot an opportunity for improvement?Share a Content Correction