missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ CSN5i-3
GREENER_CHOICE

Artikelnummer. 18709606
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18709606 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18709606 Leverandør Tocris Bioscience™ Leverandørnr. 7089/2

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Potent and selective CSN5 (COP9 signalosome) inhibitor

CSN5i-3 is a potent and selective CSN5 (COP9 signalosome) inhibitor. It inhibits deneddylation of NEDD8-modifed CRLs (IC50 = 5.8 nM), keeping them in the neddylated state, and leading to inactivation of a subset of CRLs by inducing degradation of their substrate receptor module (SRM). In A2780 ovarian cancer cells, CSN5i-3 down-regulates the expression of COPS5 and arrest cells at S-phase. In HUVECs in vitro and in zebrafish embryos in vivo, CSN5i-3 induces endothelial barrier disruption and increases macromolecule leakage by increasing the expression and activity of RhoGTPases. CSN5i-3 also inhibits the growth of lymphoma cell xenografts in mice. Orally bioavailable.

The negative control CSN5i-3-NEG (Cat. No. 7532) is also available.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 3-(Difluoromethyl)-N-(6-((5 S,6 S)-6-hydroxy-6,7,8,9-tetrahydro-5 H-imidazo[1,5-a]azepin-5-yl)-[1,10-biphenyl]-3-yl)-1-isopropyl-1 H-pyrazole-5-carboxamide
CAS 2375740-98-8
Mængde 2 mg
Mål Ubiquitin Ligase (E3) Inhibitors
Molekylær formel C28H29F2N5O2
Renhed 0.98
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.