missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CSK (Human) Recombinant Protein

Human CSK full-length ORF ( NP_004374.1, 1 a.a. - 450 a.a.) recombinant protein with GST-tag at N-terminal.

2735.00 DKK - 4145.00 DKK

Specifications

Accession Number NP_004374.1
Gene ID (Entrez) 1445
Name c-src tyrosine kinase
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16123611
View Documents
Abnova™
H00001445-P01.10ug
10 μg
2735.00 DKK
10µg
Please to purchase this item. Need a web account? Register with us today!
16133611
View Documents
Abnova™
H00001445-P01.25ug
25 μg
4145.00 DKK
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Sequence: MSAIQAAWPSGTECIAKYNFHGTAEQDLPFCKGDVLTIVAVTKDPNWYKAKNKVGREGIIPANYVQKREGVKAGTKLSLMPWFHGKITREQAERLLYPPETGLFLVRESTNYPGDYTLCVSCDGKVEHYRIMYHASKLSIDEEVYFENLMQLVEHYTSDADGLCTRLIKPKVMEGTVAAQDEFYRSGWALNMKELKLLQTIGKGEFGDVMLGDYRGNKVAVKCIKNDATAQAFLAEASVMTQLRHSNLVQLLGVIVEEKGGLYIVTEYMAKGSLVDYLRSRGRSVLGGDCLLKFSLDVCEAMEYLEGNNFVHRDLAARNVLVSEDNVAKVSDFGLTKEASSTQDTGKLPVKWTAPEALREKKFSTKSDVWSFGILLWEIYSFGRVPYPRIPLKDVVPRVEKGYKMDAPDGCPPAVYEVMKNCWHLDAAMRPSFLQLREQLEHIKTHELHL
Specifications

Specifications

NP_004374.1
c-src tyrosine kinase
125% SDS-PAGE Stained with Coomassie Blue
MGC117393
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
1445
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CSK
GST
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ CSK (Human) Recombinant Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.